Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Depth (English)0.5 in.
Depth (Metric)1.27 cm
Diameter (English) Probe0.14 in.
For Use With (Application)Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
IncludesStainless steel probe, Probe cover and Silver oxide battery
Length (English) Probe8 in.
Length (Metric)260 mm
Length (Metric) Probe20 cm
MaterialStainless Steel
ModelShockproof/Water-resistant Thermometer
Resolution0.1°C
Weight (English)0.87 oz.
Weight (Metric)24.6 g
Width (English)1.75 in.
Product TypeDigital Thermometer
Length (English)10.25 in.
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
CalibratedYes
Battery Type1.5V Silver Oxide, 357BP
Accuracy±1°C
DisplayDigital LCD
Width (Metric)4 cm
Unit SizeEach