Variable-Flow Peristaltic Pumps
Variable-Flow Peristaltic Pumps
Actual product may vary
Variable-Flow Peristaltic Pumps
Compact peristaltic pumps for use with liquids and gases
Have Questions?
Catalog NumberFlow RateElectrical Requirements
CTL13-876-20.4 to 85 mL/min.120 V 60 Hz
Catalog number CTL13-876-2
Price (MYR)
-
Flow Rate:
0.4 to 85 mL/min.
Electrical Requirements:
120 V 60 Hz
Compact peristaltic pumps for use with liquids and gases
  • Bi-directional flow with precise measured deliveries
  • Excellent flow control and flexibility for transferring and dosing liquids
  • Ultralow-, low-, medium-, and medium high-flow models feature flowrates from 0.005 to 600mL/min. and accept up to five different tubing sizes
  • Revolution of one roller delivers a precisely measured volume specific to tubing size and motor speed control
  • Variable-speed motor control and PURGE/PRIME switch for high-speed emptying and filling
  • Reverse mode permits ease in draining tubing
  • Three rollers reduce flow pulsation, prevent siphoning, and eliminate the need for check valves
  • Packaged in chemical-resistant ABS plastic case
  • Autoclavable tubing may be used with fluid temperatures from -62° to +260°C (-80° to +500°F) and is ideal for food, pharmaceutical and other critical solutions
  • Dimensions: 6.62L × 4.75W × 4.5 in.H (16.8 × 12 × 11.5cm).
Specifications
DescriptionVariable Speed Pump
Control TypeVariable Speed
Flow Rate0.4 to 85 mL/min.
Motor SpeedVariable
ReversibleYes
Includes120VAC power supply and accessory battery connector, Polypropylene fittings/nipples, Silicone tubing (0.06, 0.09, 0.18 and 0.25 in. I.D.)
Temperature (English) Max.500°F
Temperature (English) Min-80°F
Temperature (Metric) Max260°C
Temperature (Metric) Min.-62°C
Overall Length (English)6.625 in.
Overall Length (Metric)16.8 cm
Overall Width (English)4.75 in.
Overall Width (Metric)12 cm
Overall Height (English)4.5 in.
Overall Height (Metric)11.4 cm
Electrical Requirements120 V 60 Hz
Voltage120 V
Unit SizeEach