Traceable™ Handheld Digital Barometer
Traceable™ Handheld Digital Barometer
Actual product may vary
Traceable™ Handheld Digital Barometer
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
Have Questions?
Catalog NumberIncludes
CTL02-406Barometer, Lanyard, Two Button-cell Batteries, Shock-resistant Case, Battery
Catalog number CTL02-406
Price (MYR)
-
Includes:
Barometer, Lanyard, Two Button-cell Batteries, Shock-resistant Case, Battery
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
  • Stopwatch/time of day clock
  • Barometric pressure trend displays current reading and previous 2, 4, 6, 8, 10 and 12 hours in a bar chart format
  • Minimum/maximum memory for altitudes reached during any time period
  • Barometer Accuracy: ±8 mbar (0.2363 in. Hg)
Specifications
Accuracy (Barometer)±8 mbar
Resolution (Temperature)0.1°C
Temperature Range (Metric)-10°C to +60°C
Temperature Range (English)14°F to 140°F
IncludesBarometer, Lanyard, Two Button-cell Batteries, Shock-resistant Case, Battery
TypeHandheld Digital Barometer
Unit SizeEA