Traceable™ Digital Anemometer/Thermometer
Traceable™ Digital Anemometer/Thermometer
Actual product may vary
Traceable™ Digital Anemometer/Thermometer
Traceable™ Digital Anemometer/Thermometer measures outside wind speed, air velocity/temperature in hoods and slight gas movement in ducts.
Have Questions?
Catalog number CTL01-241
Price (MYR)
2,527.00
EA
Add to cart
Price (MYR)
2,527.00
EA
Add to cart
Traceable™ Digital Anemometer/Thermometer measures outside wind speed, air velocity/temperature in hoods and slight gas movement in ducts.
  • Displays temperature of air being measured; range of 32°F to 140°F and 0°C to 60°C with resolution of 0.1°F or °C
  • Designed for heavy use in lab, plant, and field with ease and accuracy
  • Instrument has four preset ranges: 0.4 to 30 mps with a resolution of 0.1 mps; 1.4 to 108 kph with a resolution of 0.1 kph; 80 to 5910 fpm with a resolution of 10 fpm; and 0.8 to 53.3 knots with a resolution of 0.1 knots
  • Anemometer accuracy is ± 2% plus 1 digit, temperature accuracy is ± 1°C
  • Low-friction, precision ball-bearing vane rotates freely in low and high velocities; reading is updated instantaneously
  • HOLD switch ''freezes'' display to capture and record readings
  • Rugged ABS case is chemical and shock resistant, and has handy flip-open bench stand for easy monitoring
  • Extra-large LCD display is 0.75 in. high (1.9 cm); can be read from 15 ft. (4.6 m)
Specifications
TypeEconomy Digital Anemometer/Thermometer
Case MaterialABS Plastic
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W)6.25 x 3 in. (15.9 x 7.6 cm)
Height (English)1.25 in.
Length (English)6.25 in.
Length (Metric)15.9 cm
Width (Metric)454 g
Temperature (Metric)Temperature (Metric)
Weight (English)16 oz.
Weight (Metric)454 g
Width (English)3 in.
Unit SizeEA