Eco Corner Sign, Rest Rooms Bilingual
Eco Corner Sign, Rest Rooms Bilingual
Actual product may vary
Eco Corner Sign, Rest Rooms Bilingual
ZING Enterprises Eco Safety sign communicates critical safety messages using a unique design that mounts easily into a wall corner.
Have Questions?
Catalog NumberMaterial
CTC2643Plastic
Catalog number CTC2643
Price (MYR)
910.00
EA
Add to cart
Material:
Plastic
Price (MYR)
910.00
EA
Add to cart
Features:
  • Alerts drivers to take precautionary measures and ensure public safety
  • Highly visible with 3M Engineer Grade Prismatic Reflective Sheeting
  • Meets MUTCD Standards
  • Made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
  • Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed
  • Recycled materials support sustainability initiatives and LEED Certified Building Projects
  • Includes pre-drilled holes for easy mounting
  • Made in the USA.
Specifications
MaterialPlastic
Quantity1 Ea.
Mounting TypeCorner Holes
For Use With (Application)Communication
Height (English)10 in.
Width (English)10 in.
LanguagesEnglish, Spanish
Dimensions (L x W)10 x 10 in.
Displayed TextRest Rooms/Banos
ColorBlue on White
Unit SizeEA