EX Head Harness
EX Head Harness
Actual product may vary
CleanSpace Respirators
EX Head Harness
EX head harness is made of TPU plastic and neoprene with nylon clips.
Have Questions?
Catalog number CSEPAF-0073
Price (MYR)
215.00
EA
Add to cart
Price (MYR)
215.00
EA
Add to cart
  • Used with the revolutionary CleanSpace™ PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Multiple adjustment points
  • Easy to wash and quick drying
  • Durable and designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Certifications/ComplianceAS/NZS1716: 2012, EN 12942 (CE Mark), Rating IP 66
DisposableReusable
Product TypeHead Harness
For Use With (Equipment)PAF-1060
ModelPAF-0073
Storage RequirementsStore at -10°C to +55°C
Weight (Metric)20 g
Unit SizeEA