Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Actual product may vary
Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsVial CapacityQuantity
CS509X52 mL Vials8120/Cs.
CS509X105 mL Vials811/Cs.
CS509X241 and 1.5 mL Vials10010/Cs.
CS509X42 mL Vials8140/Cs.
Catalog number CS509X5
Price (MYR)
4,149.00
EA
Add to cart
Holds:
2 mL Vials
Vial Capacity:
81
Quantity:
20/Cs.
Price (MYR)
4,149.00
EA
Add to cart
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
  • Warranty varies by country, please contact us for details
Specifications
TypeCryogenic Storage Box
Capacity (English)7.06 cu. ft.
Capacity (Metric)2 mL
Height (English) Exterior2 in.
Vial Capacity81
Holds2 mL Vials
Quantity20/Cs.
Unit SizeEA