Search Thermo Fisher Scientific
Catalog Number | Packaging | Material | Max. RCF |
---|---|---|---|
CRG352096 | Bulk | Polypropylene | 12,000 x g |
CRG352097 | Rack | Polypropylene | 12,000 x g |
The most dependable tube in the lab
Falcon conical tubes are easy-to-use, high quality tools to protect your precious samples. State-of-the-art design and manufacturing create tubes engineered to provide high strength, wide temperature stability, and to perform in your critical applications. They will protect your valuable samples during centrifugation, vortexing, and long term storage in the freezer. To meet this intense challenge, Falcon tubes are designed for: