L/S™ Digital Drive with Easy-Load™ 3 Pump Head for Precision Tubing
L/S™ Digital Drive with Easy-Load™ 3 Pump Head for Precision Tubing
Actual product may vary
L/S™ Digital Drive with Easy-Load™ 3 Pump Head for Precision Tubing
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easy.
Have Questions?
Catalog NumberFlow Rate
CLP77921-650.08 to 480 mL/min.
Catalog number CLP77921-65
Price (MYR)
-
Flow Rate:
0.08 to 480 mL/min.
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in mL (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
  • Models with open-head sensor stop drive motor when pump head is opened
Specifications
Certifications/ComplianceCE, cETLus, ISO 1900:2008
Channels1
Control TypeVariable Speed
Display TypeLCD
Enclosure RatingIP33
Flow Rate0.08 to 480 mL/min.
Motor Speed600 rpm
ReversibleYes
Horsepower1/10 hp
Includes07522-20 Drive, 3 m (10 ft.) Tubing, C-Flex ULTRA L/S 16 06434-16, L/S Easy-Load3 77800-60 Pump Head
Motor TypeMaintenance-free brushless motor
Pump HeadL/S Easy-Load 3 Pump Head
Overall Length (English)13.88 in.
Overall Length (Metric)352.55 mm
Overall Width (English)8.3 in.
Overall Width (Metric)210.82 mm
Overall Height (English)8.5 in.
Overall Height (Metric)215.9 mm
Electrical Requirements115/230 VAC 50/60 Hz 2.2/1.1 A
Voltage90/260 VAC
Amperage2.2/1.1 A