L/S™ Precision Tubing, C-FLEX™
L/S™ Precision Tubing, C-FLEX™
Actual product may vary
L/S™ Precision Tubing, C-FLEX™
Masterflex™ L/S™ Precision Tubing, C-FLEX™ combines biocompatibility of silicone with chemical resistance similar to Tygon™
Have Questions?
Catalog NumberDiameter (Metric) Inner
CLP06424-254.8 mm
Catalog number CLP06424-25
Price (MYR)
-
Diameter (Metric) Inner:
4.8 mm
Masterflex™ L/S™ Precision Tubing, C-FLEX™ combines biocompatibility of silicone with chemical resistance similar to Tygon™
Masterflex™ L/S™ pump tubing is manufactured to extremely close tolerances that match our L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life. Our pump tubing is factory-tested and optically inspected to provide the best performance from your peristaltic pump.
  • Very low protein binding
  • Heat sealable, weldable
  • Economical tubing
  • Applications: General-purpose lab uses requiring high performance and long life.
  • Characteristics: Exceptional tensile and tear strength; good biocompatibility; low gas permeability; and a smooth surface. Opaque white.
Specifications
Certifications/ComplianceUSP Class VI; European Pharmacopoeia (EP 3.2.9); FDA 21 CFR 177.2600; REACH, RoHS, ADCF compliant
DescriptionMasterflex™ C-Flex (C50A) tubing, L/S™ 25, 25 ft.
AutoclavableAutoclavable
Diameter (English) Inner3/16 in.
Diameter (Metric) Inner4.8 mm
MaterialC-Flex
Overall Length (English)25 ft.
Overall Length (Metric)7.6 m
For Use WithMasterflex L/S Pump Heads for Precision (Thin-Wall) Tubing
For Use With (Application)Physical properties similar to silicone with chemical compatibility of Tygon, economical, biocompatible, heat sealable and weldable
Min. Operating Temperature (English)Pumping: -40°F
SterilityNon-sterile
Tubing Size25
Flow Rate1.7 to 1000 mL/min
Hose Barb3/16 in.
Sterilization MethodEtO, autoclave, or gamma radiation. To autoclave: do not clamp, max 132°C, max 10 min.
Height (Metric)8.8 m Water
Height (English)29 ft. Water
Includes25 ft. continuous length of tubing, coiled
Gas PermeabilityO2: 200
LimitationsNot recommended for use with hydrocarbons. Moderate pumping life.
ColorOpaque White
Maximum Pressure35 psig (2.4 bar)
Vacuum Level26 in. Hg (660mm Hg)
Max. Operating Temperature (English)Pumping: 104°F
Max. Operating Temperature (Metric)Pumping: 40°C
Product TypePeristaltic Pump Tubing
Unit SizeEA