C-Flex™ L/S™ Precision Pump Tubing
C-Flex™  L/S™ Precision Pump Tubing
Actual product may vary
C-Flex™ L/S™ Precision Pump Tubing
Use this general purpose lab tubing for applications requiring high performance and long life. Masterflex™ C-Flex™ L/S™ Precision Pump Tubing combines biocompatibility of silicone with chemical resistance similar to Tygon™.
Have Questions?
Catalog NumberTubing Size
CLP06424-1717
Catalog number CLP06424-17
Price (MYR)
-
Tubing Size:
17
Use this general purpose lab tubing for applications requiring high performance and long life. Masterflex™ C-Flex™ L/S™ Precision Pump Tubing combines biocompatibility of silicone with chemical resistance similar to Tygon™.
  • Exceptional tensile and tear strength, good biocompatibility, low gas permeability and a smooth surface
  • Custom extruded to fit Masterflex™ pumps
  • Provides longer life than other platinum and peroxide silicone tubing formulations
  • ery low protein binding
  • Heat sealable, weldable
  • Better resistance to pH fluctuation
  • Complies with USP Class VI, EP, REACH, RoHS, ADCF
  • Economical
  • Opaque White
Specifications
AutoclavableAutoclavable
Diameter (Metric) Inner6.4 mm
MaterialC-Flex
Overall Length (Metric)760 cm
For Use With (Application)For general-purpose lab uses requiring high performance and long life
Min. Operating Temperature (Metric)-40°C
Tubing Size17
Flow Rate2.8 to 1700mL/min.
Hose Barb1/4 in.
Height (Metric)6.7 m
Gas PermeabilityLow
ColorOpaque White
Maximum Pressure20 psi
Vacuum Level510 mm
Max. Operating Temperature (Metric)135°C
Product TypeTubing
Unit SizeEA