Overhead Stirrer Clamp
Overhead Stirrer Clamp
Actual product may vary
Overhead Stirrer Clamp
Overhead Stirrer Clamp
Overhead Stirrer Clamp
Overhead Stirrer Clamp
Overhead Stirrer Clamp designed to accommodate stirrer support rods up to 16 mm (5/8 in.) diameter. Attaches overhead stirrer to Caframo and Fisherbrand stands (14-500-354 and 14-500-355).
Have Questions?
Catalog NumberLength (Metric)
CFMA12281 mm
Catalog number CFMA122
Price (MYR)
1,244.00
EA
Add to cart
Length (Metric):
81 mm
Price (MYR)
1,244.00
EA
Add to cart
  • This clamp is an alternative to the A120 and A120FSR (14-500-6, 14-500-357)
  • Compatible with larger diameter support rods, up to 16 mm (5/8 in.)
Specifications
For Use With (Equipment)Caframo™ and Fisherbrand™ Stands (14-500-354 and 14-500-355)
IncludesKey Only
Length (Metric)81 mm
TypeClamp
Width (Metric)40 mm
MaterialCast Zinc, Aluminum
Unit SizeEA