Shell Vials
Shell Vials
Actual product may vary
Shell Vials
Shell vials are an economical choice for many routine HPLC applications. Available in many sizes, they are sold as kits with caps to assure proper fit and function.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure TypeQuantity
C4015-96PA275 μLClearConical Insert (Polyspring)100 Pk.
C4015-96A250 μLClearConical Insert (Polyspring)Discontinued
C4011-77P2.4 mLPlug1000 Pk.
C4015-485.5 mLClearPlug100 Pk.
C4015-47P5.5 mLPlugDiscontinued
Catalog number C4015-96PA
Price (MYR)
-
Volume (Metric):
275 μL
Color:
Clear
Closure Type:
Conical Insert (Polyspring)
Quantity:
100 Pk.
Shell vials are an economical choice for many routine HPLC applications. These neckless vials with inserts are sold as kits including polyethylene push in caps to assure proper fit and function. Vial selections include clear glass, amber glass or polypropylene. 8x35mm shell vials are designed specifically for Alcott Positive Displacement autosamplers. Choose from Thermo Scientific™ National™ and Chromacol™ brands.
  • Polyethylene SepCap with starburst center design eases syringe needle penetration
  • Convenient vial kits include equal quantities of vials and caps
  • Polyspring inserts are self-aligning and provide a cushion against needle contact
  • Microsampling vials allow maximum sample extraction without need for separate inserts
Recommended for the following instruments:
  • Alcott™
  • Gilson™
  • Shimadzu™
  • Waters™ (Wisp 96 respectively 48 Position Carousel)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description300μL Polyspring Conical Insert
Diameter (Metric) Inner5 mm
ColorClear
MaterialPolypropylene
Volume (Metric)275 μL
Insert TypePolyspring Conical Insert
Volume (Metric) Residual<3μL
Base StylePrecision Point
Volume (Metric) Total275 μL
Closure TypeConical Insert (Polyspring)
Height (Metric)29 mm
Quantity100 Pk.
TypePolyspring Conical Insert
Volume (Metric) Usable250μL
Unit SizeBox