13mm Autosampler Vial Screw Thread Caps
13mm Autosampler Vial Screw Thread Caps
Actual product may vary
13mm Autosampler Vial Screw Thread Caps
13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSeptumMaterialType
C4015-1ABlackNonePolypropyleneOpen Top Screw Cap
12-SCSBlackNone (no hole)PolypropyleneScrew Cap
Catalog number C4015-1A
Price (MYR)
-
Color:
Black
Septum:
None
Material:
Polypropylene
Type:
Open Top Screw Cap
Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Autosampler Vial Screw Thread Caps
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionBlack; Polypropylene
ColorBlack
MaterialPolypropylene
SeptumNone
Quantity100
TypeOpen Top Screw Cap
Closure Size13 mm
For Use With (Equipment)13-425 Screw Thread Vials
Unit SizePack