8mm Plastic Screw Thread Vials
8mm Plastic Screw Thread Vials
Actual product may vary
8mm Plastic Screw Thread Vials
Choose from these chemically inert polypropylene vials that are suitable for most chromatography applications.
Have Questions?
Catalog NumberVolume (Metric)
C4013-11475 μL
Catalog number C4013-11
Price (MYR)
-
Volume (Metric):
475 μL
Choose from these economical alternanatives to glass vials. These 8mm plastic standard opening screw thread vials are manufactured from high-quality, GC-tested polypropylene material and feature an 8-425 finish. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Broad application of polypropylene vials
  • Chemically inert and suitable for most chromatography applications.
  • Preferable in some cases when levels of inorganic ions have to be kept to an absolute minimum
  • Economical alternative to glass
Chemically resistant
  • The plastic material of choice when storing liquid sample in aqueous/organic mixtures due to its wide chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
  • Tolerates temperatures from -80°C to +121°C
Autosampler compatibility*
Beckman™
CTC™
Gilson™
Knauer™
Shimadzu™
Spark Holland™
Varian™
VWR™ (Merck)/Hitachi™
*See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description250µL Conical MicroVial; Polypropylene
Diameter (Metric) Opening8 mm
Diameter (Metric) Inner12 mm
ColorClear
Diameter (Metric) Outer12 mm
MaterialPolypropylene
Volume (Metric)475 μL
Insert Type250μL Conical MicroVial
Volume (Metric) Residual<4μL
Volume (Metric) Working250 μL
Base StyleConical
Volume (Metric) Total475 μL
Closure TypeScrew Cap
Height (Metric)32 mm
Quantity100
TypeScrew Thread Vial
Closure Size8-425
Marking SpotNo
Volume (Metric) Usable250μL
Unit SizeBox