8mm Autosampler Inserts
8mm Autosampler Inserts
Actual product may vary
8mm Autosampler Inserts
Clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 125μL to 200μL.
Have Questions?
Catalog NumberVolume (Metric)MaterialBase StyleQuantity
C4012-529200 μLGlassConical100
Catalog number C4012-529
Price (MYR)
-
Volume (Metric):
200 μL
Material:
Glass
Base Style:
Conical
Quantity:
100
These clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125-200μL.
  • Pulled point inserts are an economical choice for non-critical applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description150µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
StylePulled Point
Volume (Metric) Residual<3 μL
Base StyleConical
Volume (Metric) Total200 μL
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Vial Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack