Manual Crimpers and Decrimpers for Cleanrooms
Manual Crimpers and Decrimpers for Cleanrooms
Actual product may vary
Manual Crimpers and Decrimpers for Cleanrooms
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeType
C4012-102SS11 mmManual Decrimper
C4013-100SS13 mmManual Crimper
Catalog number C4012-102SS
Price (MYR)
-
Size:
11 mm
Type:
Manual Decrimper
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
No. per Pack1 Ea.
TypeManual Decrimper
For Use With (Equipment)Aluminum Seal
Size11 mm
MaterialStainless Steel
For Use With (Application)Industrial Chromatography
Product LineNational
Unit SizeEach