9mm Autosampler Inserts
9mm Autosampler Inserts
Actual product may vary
9mm Autosampler Inserts
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
Have Questions?
Catalog NumberVolume (Metric)MaterialBase StyleInsert Type
C4010-630375 μLGlassConicalPolyspring Insert
Catalog number C4010-630
Price (MYR)
927.00
PK
Add to cart
Volume (Metric):
375 μL
Material:
Glass
Base Style:
Conical
Insert Type:
Polyspring Insert
Price (MYR)
927.00
PK
Add to cart
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
  • Pulled point inserts are an economical choice for noncritical applications
  • Nominal O.D. of 6mm to fit Target wide-opening vials
For use with:
  • Target LoVial Wide-Opening vials
  • Snap-It™ Wide-Opening Snap Cap vials
  • Target DP™ vials
  • Target Wide-Opening vials
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description300µL Polyspring Insert
Diameter (Metric) Inner6 mm
ColorClear
Diameter (Metric) Outer6 mm
MaterialGlass
Volume (Metric)375 μL
Insert TypePolyspring Insert
StylePolyspring centers and cushions insert
Volume (Metric) Residual<1 μL
Base StyleConical
Volume (Metric) Total375 μL
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Vial Insert
Volume (Metric) Usable300 μL
For Use With (Equipment)National 11 mm Crimp Top Vials
Unit SizePK