Search Thermo Fisher Scientific
Catalog Number | Type | Includes |
---|---|---|
C4000-MS | Empty Mini-Storeroom Cabinet | Cabinet Only |
Organize your supplies and save valuable bench space with Thermo Scientific™ National™ 8mm Standard Opening Screw Thread Vial Storerooms. Storerooms are available in six and nine drawer models. The six drawer mini-storeroom holds 500 vials and closures; the nine drawer full size storeroom holds 2000 vials and closures. Order an empty cabinet or one fully stocked with vials and closures.