9mm Autosampler Vial Septa
9mm Autosampler Vial Septa
Actual product may vary
9mm Autosampler Vial Septa
9mm Septa are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE to suit a variety of applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorMaterialHardness
C4000-30Ivory, RedPTFE, Rubber35 °shore
C4000-40Red, WhitePTFE, Silicone45 °shore
Catalog number C4000-30
Price (MYR)
-
Color:
Ivory, Red
Material:
PTFE, Rubber
Hardness:
35 °shore
Thermo Scientific™ 9mm Septa are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE to suit a variety of applications. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSeptum for 9mm Screw Thread Cap
ColorIvory, Red
MaterialPTFE, Rubber
SeptumSilicone/PTFE
Hardness35 °shore
Quantity100
Thickness (Metric)1.0 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size9 mm
For Use With (Equipment)9 mm Screw Thread Cap
Unit SizePack