Filter Funnel
Filter Funnel
Actual product may vary
Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
Have Questions?
Catalog NumberCapacity (Metric)Pore Size
BSN21255250 mL5
Catalog number BSN21255
Price (MYR)
3,206.00
EA
Add to cart
Capacity (Metric):
250 mL
Pore Size:
5
Price (MYR)
3,206.00
EA
Add to cart
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass (3.3)
DescriptionBuchner Shape
Pore Size5
Unit SizeEA