ValuSeal™ 1500 Wipers
ValuSeal™ 1500 Wipers
Actual product may vary
ValuSeal™ 1500 Wipers
For ISO Class 3 (Class 1) applications
Have Questions?
Catalog NumberDimensions (L x W)Description
BERSPXV1500.004.1212 x 4 in. (30.4 x 10.1 cm)SatPax™
Catalog number BERSPXV1500.004.12
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
12 x 4 in. (30.4 x 10.1 cm)
Description:
SatPax™
  • Strong, laundered, 100% knit polyester is tear- and abrasion-resistant
  • Economical laser-cut, sealed-edge wipers provide high performance at a low cost
  • Sealed edges minimize the potential of contamination from particles, fibers, ions, and residue
  • Works well with IPA, acetone and other solvents
  • Double-knit construction makes wipes extremely absorbent—more cleanup with fewer wipers for reduced costs and waste
  • Steam autoclavable for aseptic environments
Specifications
Product TypePre-saturated Wipe
Length (English)12 in.
Length (Metric)30.4 cm
Width (English)4 in.
Width (Metric)10.1 cm
MaterialPolyester Knit
AutoclavableAutoclavable
Basis Weight120
Certifications/ComplianceOSHA
ColorWhite
ConstructionDouble-knit
DescriptionSatPax™
Dimensions (L x W)12 x 4 in. (30.4 x 10.1 cm)
For Use With (Application)For ISO Class 3 (Class 1) applications
Unit SizeCS