Components for Vial Convenience Kits
Components for Vial Convenience Kits
Actual product may vary
Components for Vial Convenience Kits
Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.
Have Questions?
Catalog NumberVolume (Metric)Color
B7921-VO20 mLAmber
Catalog number B7921-VO
Price (MYR)
-
Volume (Metric):
20 mL
Color:
Amber
Vials, caps and septa for use with Thermo Scientific™ National vial convenience kits.
Specifications
DescriptionAmber 20mL vials, 28 x 57mm
ColorAmber
Diameter (Metric) Outer28 mm
Volume (Metric)20 mL
Product LineNational
Volume (Metric) Total20 mL
Closure TypeScrew Cap
Height (Metric)57 mm
Quantity100 Pk.
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
For Use With (Equipment)Thermo Scientific™ National Vial Convenience Kits
Unit SizeEach