Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Actual product may vary
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Owl™ EasyCast™ B3 Gel System with Built-In Recirculation
Provide uniform, reproducible results with the Thermo Scientific™ Owl™ EasyCast™ B3 Gel System with Built-in Recirculation, ideal for long runs or RNA gels.
Have Questions?
Catalog NumberGel Size
B312 x 14 cm
Catalog number B3
Price (MYR)
5,824.00
EA
Add to cart
Gel Size:
12 x 14 cm
Price (MYR)
5,824.00
EA
Add to cart
Provide uniform, reproducible results with the Thermo Scientific™ Owl™ EasyCast™ B3 Gel System with Built-in Recirculation, ideal for long runs, multiple sample sets or RNA gels.
Features:
  • Prevents the formation of pH and ionic gradients
  • Delivers clear results for samples run over long time periods
  • Requires no external pumps, tubing or stir bars, as recirculation system is built into buffer chamber
  • Eliminates uneven migration, band distortion or disassociation of pH-dependent glyoxylated RNA molecules that can result when ionic depletion occurs
Includes:
  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ gasketed UVT gel tray
  • 12- and 20-well, double-sided 1.0/1.5mm thick combs
Specifications
Certifications/ComplianceCE marked
DescriptionEasyCast; Gel Size: 12 x 14cmL; Buffer volume: 1L; L x W x H: 25 x 18 x 12cm
Gel Size12 x 14 cm
Product LineOwl
TypeMini Gel Electrophoresis System
Sample Size8 to 50
Volume (Metric) Buffer1000 mL
Dimensions (L x W x H)9.75 x 7 x 4.72 in. (25 x 18 x 12 cm)
Length (English)9.75 in.
Length (Metric)25 cm
Length (Metric) Gel14 cm
Width (English)7 in.
Width (Metric)18 cm
Width (Metric) Gel12 cm
Height (English)4.72 in.
Height (Metric)12 cm
Unit SizeEA
Frequently asked questions (FAQs)
Is the PowerEase Touch Power Supply (Cat. No. PS0351) compatible with the Owl gel Systems?
Yes, it is compatible with the Owl Gel Systems. The following ones can be used with the PowerEase power supplies with no need for adaptors:
    Owl EasyCast B1 Mini Gel Electrophor. Sys. (Cat. No. B1-BP)
    Owl A2 Large Gel Sys. (Cat. No. A2)
    Owl EasyCast B3 Gel Sys. with Built-In Recirculat. (Cat. No. B3).
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.