Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Actual product may vary
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.
Have Questions?
Catalog NumberIncludes
B2EasyCast
Catalog number B2
Price (MYR)
4,396.00
EA
Add to cart
Includes:
EasyCast
Price (MYR)
4,396.00
EA
Add to cart
Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.
Features:
  • Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed U.V. Transmissible (UVT) gel tray
  • Two combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
Specifications
Certifications/ComplianceCE marked
Gel SizeMini
Product LineOwl
TypeMini Gel Electrophoresis System
IncludesEasyCast
Sample Size8 to 48
Volume (Metric) Buffer800 mL
Dimensions (L x W x H)9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm)
Length (English)9.63 in.
Length (Metric)24.5 cm
Length (Metric) Gel14 cm
Width (English)7.06 in.
Width (Metric)18 cm
Width (Metric) Gel12 cm
Height (English)3.75 in.
Height (Metric)9.5 cm
Max. Voltage150 V
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.