Check Order Status
Nalgene™ System 100™ Cryogenic Vials
Actual product may vary
Nalgene™ System 100™ Cryogenic Vials
Externally threaded polypropylene self-standing vials
Have Questions?
Change view
Catalog Number
Volume (Metric)
AY509X33
1.5 mL
AY509X32
1 mL
2 Options
Catalog number
AY509X33
HSN
84189900
Price (MYR)
4,069.00
CS
Add to cart
Volume (Metric):
1.5 mL
Price (MYR)
4,069.00
CS
Add to cart
Sterile
For use with System 100 CryoBoxes™ and 25- and 81-ampule boxes with 2.0mL ampule capacities
Specifications
Material
Polypropylene
Sterility
Sterile
Volume (Metric)
1.5 mL
Thread Style
External
Quantity
500/Cs.