Nalgene™ System 100™ Cryogenic Vials
Nalgene™ System 100™ Cryogenic Vials
Actual product may vary
Nalgene™ System 100™ Cryogenic Vials
Externally threaded polypropylene self-standing vials
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
AY509X321 mL
AY509X331.5 mL
Catalog number AY509X32
Price (MYR)
4,069.00
EA
Add to cart
Volume (Metric):
1 mL
Price (MYR)
4,069.00
EA
Add to cart
  • Sterile
  • For use with System 100 CryoBoxes™ and 25- and 81-ampule boxes with 2.0mL ampule capacities
Specifications
MaterialPolypropylene
SterilitySterile
Volume (Metric)1 mL
Thread StyleExternal
Quantity500/Cs.
Unit SizeEA