Cerium(IV) sulfate, anhydrous, 97%
Cerium(IV) sulfate, anhydrous, 97%
Cerium(IV) sulfate, anhydrous, 97%
Cerium(IV) sulfate, anhydrous, 97%
Thermo Scientific Chemicals
Cerium(IV) sulfate, anhydrous, 97%
CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFL20254.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) sulfate
Name Noteanhydrous
CAS13590-82-4
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
View more
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions' reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cerium(IV) sulfate is a redox indicator and is used in analytical chemistry for redox titrations. In ionized form, ceric ion acts as a strong oxidizer under acidic conditions. It is also used to prepare spherical-like nanoparticles CeO2, nanoparticles for NO reductions′ reactions.
Solubility
Soluble in water and dilute sulfuric acid.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Avoid moisture. Incompatible with strong reducing agents.
RUO – Research Use Only
General References:
  1. Khan, M. M. A.; Naushad, M. Synthesis, surface characterization and electrochemical properties of PVC-based cerium(IV) sulphate ion exchange composite membrane. Ionics 2015, 21 (4), 1057-1062.
  2. Asutay, O.; Hamarat, N.; Uludag, N.; Coskun, N. Selective oxidative deoximation with anhydrous Ce(IV) sulfate. Tetrahedron Lett. 2015, 56 (25), 3902-3904.