4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%, Thermo Scientific™
4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%, Thermo Scientific™
4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%, Thermo Scientific™
4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%, Thermo Scientific™
4-Methyl-2,3,5,6-tetrafluorobenzyl bromide, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFL20210.14
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Methyl-2,3,5,6-tetrafluorobenzyl bromide
CAS92814-00-1
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.