3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
3-Amino-2-bromopyridine, 97%
Thermo Scientific Chemicals
3-Amino-2-bromopyridine, 97%
CAS: 39856-58-1 | C5H5BrN2 | 173.013 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19839.1425 g
Catalog number ALFL19839.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Amino-2-bromopyridine
CAS39856-58-1
Health Hazard 1H301+H311+H331-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
View more
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Mohamed Shaker S. Adam,; Olaf Kühl,; Markus K. Kindermann,; Joachim W. Heinicke,; Peter G. Jones. 3-Amino- and 3-acylamido-2-phosphonopyridines: synthesis by Pd-catalyzed P-C coupling, structure and conversion to pyrido[b]-anellated Pdouble bond; length as m-dashC-N heterocycles. Tetrahedron. 2008, 64(34), 7960-7967.