2-Amino-6-chloropyridine, 98%
2-Amino-6-chloropyridine, 98%
2-Amino-6-chloropyridine, 98%
2-Amino-6-chloropyridine, 98%
Thermo Scientific Chemicals
2-Amino-6-chloropyridine, 98%
CAS: 45644-21-1 | C5H5ClN2 | 128.559 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19836.065 g
Catalog number ALFL19836.06
Price (MYR)
622.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
622.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-6-chloropyridine
CAS45644-21-1
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c