1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%
CAS: 155371-19-0 | C6H11F6N2P | 256.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
Catalog number ALFL19762.18
Price (MYR)
2,482.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
2,482.00
EA
Add to cart
Specifications
Chemical Name or Material1-Ethyl-3-methylimidazolium hexafluorophosphate
CAS155371-19-0
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362