4-(Methylamino)pyridine, 99%
4-(Methylamino)pyridine, 99%
4-(Methylamino)pyridine, 99%
4-(Methylamino)pyridine, 99%
Thermo Scientific Chemicals
4-(Methylamino)pyridine, 99%
CAS: 1121-58-0 | C6H8N2 | 108.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19626.1425 g
Catalog number ALFL19626.14
Price (MYR)
2,592.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,592.00
EA
Add to cart
Specifications
Chemical Name or Material4-(Methylamino)pyridine
CAS1121-58-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.
Solubility
Soluble in water, acetone, ethanol, ether and benzene.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Kin-Shan Huang.; Doyle Britton.; Stephen R. Byrn. A novel class of phenol-pyridine co-crystals for secondharmonic generation. Journal of Materials Chemistry. 1997, 7 (5), 713-720.
  2. Irena Pulko Dr.; Jennifer Wall.; Peter Krajnc Prof.; Neil R. Cameron Prof. Ultra-High Surface Area Functional Porous Polymers by Emulsion Templating and Hypercrosslinking: Efficient Nucleophilic Catalyst Supports. Chemistry - A European Journal. 2010, 16 (8), 2350-2354.