N-Boc-ethylenediamine hydrochloride, 98%
N-Boc-ethylenediamine hydrochloride, 98%
N-Boc-ethylenediamine hydrochloride, 98%
N-Boc-ethylenediamine hydrochloride, 98%
Thermo Scientific Chemicals
N-Boc-ethylenediamine hydrochloride, 98%
CAS: 79513-35-2 | C7H17ClN2O2 | 196.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL19485.06
Price (MYR)
2,515.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,515.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Boc-ethylenediamine hydrochloride
Melting Point148°C to 151°C
CAS79513-35-2
Recommended StorageAmbient temperatures; Store under Nitrogen
EINECS Number000-000-0