3-Fluorosalicylaldehyde, 98%
3-Fluorosalicylaldehyde, 98%
3-Fluorosalicylaldehyde, 98%
3-Fluorosalicylaldehyde, 98%
Thermo Scientific Chemicals
3-Fluorosalicylaldehyde, 98%
CAS: 394-50-3 | C7H5FO2 | 140.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19324.065 g
Catalog number ALFL19324.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Fluorosalicylaldehyde
CAS394-50-3
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c