Glyoxal dimethyl acetal, 60% aq. soln.
Glyoxal dimethyl acetal, 60% aq. soln.
Glyoxal dimethyl acetal, 60% aq. soln.
Glyoxal dimethyl acetal, 60% aq. soln.
Thermo Scientific Chemicals
Glyoxal dimethyl acetal, 60% aq. soln.
CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFL18490.36
Price (MYR)
1,989.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,989.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlyoxal dimethyl acetal
CAS51673-84-8
Health Hazard 1H317
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c
View more
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Pacsu, E.; Trister, S. M.; Green, J. W. The Acetone Derivatives of the Mercaptals of Some Monosaccharides. IV. 1 The 4, 5-Monoacetone Derivative of the Dibenzylmercaptal and of the Dimethylacetal of d-Galactose. Journal of the American Chemical Society. 1939, 61 (9), 2444-2448.
  2. Laurent Soulère.; Yves Queneau.; Alain Doutheau. Kissick. An expeditious synthesis of 4-hydroxy-2(E)-nonenal (4-HNE), its dimethyl acetal and of related compounds. Chemistry and Physics of Lipids. 2007, 150 (2), 239-243.