N-(5-Bromopentyl)phthalimide, 97%
N-(5-Bromopentyl)phthalimide, 97%
N-(5-Bromopentyl)phthalimide, 97%
N-(5-Bromopentyl)phthalimide, 97%
Thermo Scientific Chemicals
N-(5-Bromopentyl)phthalimide, 97%
CAS: 954-81-4 | C13H14BrNO2 | 296.164 g/mol
Have Questions?
Catalog NumberQuantity
ALFL17543.1425 g
Catalog number ALFL17543.14
Price (MYR)
881.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
881.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-(5-Bromopentyl)phthalimide
CAS954-81-4
Melting Point58°C to 62°C
Recommended StorageAmbient temperatures
TSCANo
View more
It is a pharmaceutical intermediate and is used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate and is used in medicine.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Richard A. Glennon.; Noreen A. Naiman.; Robert A. Lyon.; Milt Titeler. Arylpiperazine derivatives as high-affinity 5-HT1A serotonin ligands.J. Med. Chem. 1988, 31 (10),1968-1971 .
  2. Richard A. Glennon.; Noreen A. Naiman.; M. Edward Pierson.; J. Doyle Smith.; Abd M. Ismaiel.; Milt Titeler.; Robert A. Lyon. N-(phthalimidoalkyl) derivatives of serotonergic agents: a common interaction at 5-HT1A serotonin binding sites?J. Med. Chem. 1989, 32 (8),1921-1926 .