Dimethyl azelate, 97%
Dimethyl azelate, 97%
Dimethyl azelate, 97%
Dimethyl azelate, 97%
Thermo Scientific Chemicals
Dimethyl azelate, 97%
CAS: 1732-10-1 | C11H20O4 | 216.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFL17458.22100 g
Catalog number ALFL17458.22
Price (MYR)
2,422.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,422.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl azelate
CAS1732-10-1
Melting Point18°C
Recommended StorageAmbient temperatures
Density1.007