Formic acid, 85%
Formic acid, 85%
Formic acid, 85%
Thermo Scientific Chemicals
Formic acid, 85%
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
1000 g
Catalog number ALFL17434.0E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFormic acid
CAS64-18-6
Melting Point7°C to 9°C
Boiling Point100°C to 101°C
Health Hazard 1H226-H302-H314-H331
View more
Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Formic acid is principally used as a preservative antibacterial agent in livestock feed, in dying textiles, tanning leather and electroplating. In the poultry industry, it is sometimes added with feed to kill E. coli bacteria. It acts as a reducing agent for the catalytic reduction of nitrate in water.
Solubility
Miscible with water, ether, acetone, ethyl acetate, glycerol, methanol and ethanol. Partially miscible with benzene, toluene and xylene.
Notes
Hygroscopic. Store closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only
General References:
  1. Griggs, J. P. Alternatives to Antibiotics for Organic Poultry Production. J. Appl. Poult. Res. 2005, 14 (4), 750-756.
  2. Garron, A.; Epron, F. Use of formic acid as reducing agent for application in catalytic reduction of nitrate in water. Water Res. 2005, 39 (13), 3073-3081.