Silicon(IV) oxide, powder, 0.5 micron, 99.9%
Silicon(IV) oxide, powder, 0.5 micron, 99.9%
Silicon(IV) oxide, powder, 0.5 micron, 99.9%
Silicon(IV) oxide, powder, 0.5 micron, 99.9%
Thermo Scientific Chemicals
Silicon(IV) oxide, powder, 0.5 micron, 99.9%
Silicon(IV) oxide, 99.9%, O2Si, CAS Number-7631-86-9, silica, amorphous, silicic anhydride, tridymite, infusorial earth, silicon dioxide, silica, cristobalite, quartz, aerosil, diatomaceous earth, 100g, 2230 deg.C, CHEBI:30563, 231-545-4, 60.09, Warning | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16985.22100 g
Catalog number ALFL16985.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon(IV) oxide
Melting Point1710°C
Name Note0.5 micron
CAS7631-86-9
View more
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
Solubility
Insoluble in water.
Notes
It is a very stable compound of silicon.
RUO – Research Use Only
General References:
  1. High purity, spherical silica powder of narrow particle size distribution.
  2. Ojovan, M. I. Glass formation in amorphous SiO2 as a percolation phase transition in a system of network defects. J. Exp. Theor. Phys. Lett. 2004, 79 (12), 632-634.
  3. Nandiyanto, A. B. D.; Kim, S. G.; Iskandar, F.; Okuyama, K. Synthesis of spherical mesoporous silica nanoparticles with nanometer-size controllable pores and outer diameters. Micropor. Mesopor. Mat. 2009, 120 (3), 447.