n-Decyltrimethoxysilane, 97%
n-Decyltrimethoxysilane, 97%
n-Decyltrimethoxysilane, 97%
n-Decyltrimethoxysilane, 97%
Thermo Scientific Chemicals
n-Decyltrimethoxysilane, 97%
CAS: 5575-48-4 | C13H30O3Si | 262.47 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16966.1425 g
Catalog number ALFL16966.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point102°C to 106°C (3.5mmHg)
Chemical Name or Materialn-Decyltrimethoxysilane
Melting Point-37°C
CAS5575-48-4
Health Hazard 1H315-H319
View more
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Jing He.; Yan Xu.; Hui Ma.; Qiong Zhang.; David G. Evans.; Xue Duan. Effect of surface hydrophobicity/hydrophilicity of mesoporous supports on the activity of immobilized lipase. Journal of Colloid and Interface Science. 2006, 298 (2), 780-786.
  2. Marianna Kemell.; Elina Färm.; Markku Leskelä.; Mikko Ritala. Transparent superhydrophobic surfaces by self-assembly of hydrophobic monolayers on nanostructured surfaces. physica status solidi (a). 2006, 203 (6), 1453-1458.