Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%
Thermo Scientific Chemicals
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%
CAS: 28523-86-6 | C4H3F7O | 200.056 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL16891.06
Price (MYR)
1,677.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,677.00
EA
Add to cart
Specifications
Chemical Name or MaterialFluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether
CAS28523-86-6
Recommended StorageAmbient temperatures
Density1.505
RTECS NumberKO0737000
View more
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kawada, C.; Kimura, M.; Masuda, Y.; Nomura, Y. Oral administration of hyaluronan prevents skin dryness and epidermal thickening in ultraviolet irradiated hairless mice. J. Photochem. Photobiol., B 2015, 153, 215-221.
  2. Munekage, M.; Yatabe, T.; Kitagawa, H.; Takezaki, Y.; Tamura, T.; Namikawa, T.; Hanazaki, K. An artificial pancreas provided a novel model of blood glucose level variability in beagles. J. Artif. Organs 2015, 18 (4), 387-390.