1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFL16585.065 g
Catalog number ALFL16585.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Boiling Point103°C to 106°C (3mmHg)
Chemical Name or Material1H,1H,2H,2H-Perfluorodecyltriethoxysilane
CAS101947-16-4
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1H,1H,2H,2H-Perfluorodecyltriethoxysilane is used to bond poly(tetrafluoroethylene) films to silicon wafers. It is also utilized in anti-reflective coating, release coating and soil repellent coating.
Solubility
Miscible with ethanol and tetrahydrofuran.
Notes
Incompatible with strong oxidizing agents.
General References:
  1. Has been used to bond poly(tetrafluoroethylene) films to silicon wafers.
  2. Meng, S.; Ye, Y.; Mansouri, J.; Chen, V. Fouling and crystallisation behaviour of superhydrophobic nano-composite PVDF membranes in direct contact membrane distillation. J. Membr. Sci. 2014, 463, 102-112.
  3. Meng, S.; Ye, Y.; Mansouri, J.; Chen, V. Crystallization behavior of salts during membrane distillation with hydrophobic and superhydrophobic capillary membranes. J. Membr. Sci. 2015, 473, 165-176.