(3-Aminopropyl)diethoxymethylsilane, 97%
(3-Aminopropyl)diethoxymethylsilane, 97%
(3-Aminopropyl)diethoxymethylsilane, 97%
(3-Aminopropyl)diethoxymethylsilane, 97%
Thermo Scientific Chemicals
(3-Aminopropyl)diethoxymethylsilane, 97%
CAS: 3179-76-8 | C8H21NO2Si | 191.346 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16401.1425 g
Catalog number ALFL16401.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point197°C to 199°C
Chemical Name or Material(3-Aminopropyl)diethoxymethylsilane
Melting Point<-20°C
CAS3179-76-8
Health Hazard 1H227-H314-H335
View more
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.
Solubility
Soluble and reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Young-Hye La,; Hyun Ju Kim,; Il Sang Maeng,; Yu Jin Jung,;Joon Won Park. Selective Cleavage of Nitro Groups in Nitro-Substituted Aromatic Monolayers by Synchrotron Soft X-rays:  Effect of Molecular Structure on Cleavage Rates. Langmuir. 2002, 18 (6), 2430-2433.