Dimethoxymethylsilane, 97%
Dimethoxymethylsilane, 97%
Dimethoxymethylsilane, 97%
Dimethoxymethylsilane, 97%
Dimethoxymethylsilane, 97%
CAS: 16881-77-9 | C3H9O2Si | 105.188 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16197.1425 g
Catalog number ALFL16197.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point61°C
Chemical Name or MaterialDimethoxymethylsilane
Melting Point-136°C
CAS16881-77-9
Health Hazard 1H225