1-Vinylimidazole, 99%
1-Vinylimidazole, 99%
1-Vinylimidazole, 99%
1-Vinylimidazole, 99%
Thermo Scientific Chemicals
1-Vinylimidazole, 99%
CAS: 1072-63-5 | C5H6N2 | 94.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL16174.22
Price (MYR)
462.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
462.00
EA
Add to cart
Specifications
Chemical Name or Material1-Vinylimidazole
CAS1072-63-5
Health Hazard 1H227-H302-H318-H360D
Health Hazard 2GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Health Hazard 3P201-P202-P210-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
View more
1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.
Solubility
Miscible with water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Hamilton, J. R.; Abedini, A.; Zhang, Z.; Whitley, J. W.; Bara, J. E.; Turner, C. H. Enhancing the pre-polymerization coordination of 1-vinylimidazole. Chem. Eng. Sci. 2015, 138, 646-654.
  2. Primo, G. A.; Manzano, M. F. G.; Romero, M. R.; Igarzabal, C. I. A. Synthesis and characterization of hydrogels from 1-vinylimidazole. Highly resistant co-polymers with synergistic effect. Mater. Chem. Phys. 2015, 153, 365-375.