Di-n-propylamine, 99%
Di-n-propylamine, 99%
Di-n-propylamine, 99%
Di-n-propylamine, 99%
Thermo Scientific Chemicals
Di-n-propylamine, 99%
CAS: 142-84-7 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL15808.AP
Price (MYR)
388.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
388.00
EA
Add to cart
Specifications
Chemical Name or MaterialDi-n-propylamine
CAS142-84-7
Health Hazard 1H225-H302-H311+H331-H314-H335
Health Hazard 2GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c