Polysorbate 20
Polysorbate 20
Polysorbate 20
Thermo Scientific Chemicals
Polysorbate 20
CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15029.AE100 mL
Catalog number ALFL15029.AE
Price (MYR)
243.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
243.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolysorbate 20
CAS9005-64-5
Boiling Point>100°C
Recommended StorageAmbient temperatures
RTECS NumberTR7400000
View more
Used for solubilizing membrane proteins during isolation and purification.Polysorbate 20 is used as a wetting agent in flavored mouth drops. It acts as a washing agent in Western blots and ELISAs and prevents antibody binding. It is a solubilizing agent of membrane proteins. In pharmaceutical chemistry, it is used as an excipient to stabilize emulsions and suspensions. Further, it is used as a dispersing agent, stabilizer, lubricator and also used in creams, salves, ointments, protective creams, balms, pomades, lipsticks, mascaras and glosses.