Polypropylene glycol 2000
Polypropylene glycol 2000
Polypropylene glycol 2000
Polypropylene glycol 2000
Thermo Scientific Chemicals
Polypropylene glycol 2000
CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL14699.AP
Price (MYR)
587.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
587.00
EA
Add to cart
Specifications
Chemical Name or MaterialPolypropylene glycol 2000
CAS25322-69-4
Recommended StorageAmbient temperatures
RTECS NumberTR6130000
EINECS Number500-039-8
View more
Used to make polyester compounds, absorb extra water and maintain moisture in certain medicines, cosmetics, or food products and as a base for deicing solutions. Propylene glycol is used by the chemical, food, and pharmaceutical industries as an antifreeze. It is a solvent for food colors and flavors, and in the paint and plastics industries. Propylene glycol is also used to create artificial smoke or fog used in fire-fighting training and in theatrical productions.