Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol
Thermo Scientific Chemicals
Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol
CAS: 78-79-5 | C5H8 | 68.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL14619.AP
Price (MYR)
528.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
528.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsoprene
Name Notestabilized with ≈0.02% 4-tert-butylcatechol
CAS78-79-5
Health Hazard 1H224-H341-H350
Health Hazard 2GHS H Statement
H224-H350-H341
Extremely flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
View more
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.
Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone and benzene. Immiscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, ammonia, alkali metals, alcohols and oxygen.
RUO – Research Use Only
General References:
  1. Kuwata, M.; Liu, Y.; McKinney, K.; Martin, S. T. Physical state and acidity of inorganic sulfate can regulate the production of secondary organic material from isoprene photooxidation products. Phys. Chem. Chem. Phys. 2015, 17 (8), 5670-5678.
  2. Loreto, F.; Fineschi, S. Reconciling functions and evolution of isoprene emission in higher plants. New Phytol. 2015, 206 (2), 578-582.